Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Rabbit anti-Human, Mouse, Rat, Clone: SN0646, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267665
Description
PDGF R beta Monoclonal antibody specifically detects PDGF R beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
PDGFRbeta | |
SN0646 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Purified | |
PDGFRB | |
Synthetic phospho-peptide corresponding to residues surrounding Tyrosine 740 of Human PDGF Receptor beta | |
Protein A purified | |
Oncogenes, Phospho Specific, Protein Kinase | |
Primary | |
5159 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title