Learn More
Aviva Systems Biology Corp PDHA1 Antibody - C-terminal region (ARP48137_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48137_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PDHA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PDHA1. Alternate names: PDHA, PDHCE1A, PHE1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59724