Learn More
Enzo Life Sciences PDI polyclonal antibody (50 µg)
Supplier: Enzo Life Sciences ADISPA890D
Host: Rabbit. Immunogen: Purified bovine liver Protein Disulfide Isomerase. UniProt: P05307. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: ICC, IHC (PS), IP, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.