Learn More
Enzo Life SciencesSupplier Diversity Partner IMMUNOSET PDI ELISA development set (96 wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI960072
The IMMUNOSET PDI ELISA development set contains sufficient reagents for the development of 5 x 96 well ELISA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own ELISA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules. Alternative name: Protein disulfide isomerase. P4HB. Contents: Capture antibody, Standard, Detection antibody, SA-HRP. Application: ELISA, Colorimetric detection. Assay Time: Plate Coating - Overnight + 1 hour; Assay - 3 hours. Crossreactivity: PDI (100%), Grp94 (0.121%), Hsc70 (0.018%) and 0.016%: Calreticulin, Grp78, HSP90α, HSP90β, and HSP70. Long Term Storage: +4°C
Catalog No. 50-658-676