PDI™ Prevantics™ Antiseptic Products

The first FDA-approved 3.15% chlorhexidine gluconate and 70% isopropyl alcohol skin antiseptic

Manufacturer: Nice Pak ProductsSupplier Diversity Partner B11400

 View more versions of this product

Catalog No. 23-100-113

  Check Availability
Add to Cart

    Specifications

    Size (English) 0.17 oz.
    Size (Metric) 5.1mL
    Quantity 3000 Case
    Certifications/Compliance FDA
    Composition Chlorhexidine 3.15% w/v, Isopropyl Alcohol 70% v/v
    For Use With (Application) For pre-operative or pre-injection skin antisepsis
    Product Type Antiseptic Skin Swabstick

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit