Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDI Rabbit anti-Human, Clone: JF97-08, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267030

Catalog No. NBP267030


Add to cart

Description

Description

PDI Monoclonal antibody specifically detects PDI in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

PDI
JF97-08
Unconjugated
Purified
Pancreas-specific protein disulfide isomerase, pancreatic protein disulfide isomerase, PDA2, PDI, PDIp, PDIPEC 5.3.4.1, PDIR, protein disulfide isomerase A2, protein disulfide isomerase family A, member 2, protein disulfide isomerase, pancreatic, protein disulfide isomerase-associated 2, protein disulfide-isomerase A2, Rho GDP dissociation inhibitor gamma
PDIA2
Recombinant Protein
Monoclonal
Protein A purified
Biologically Active Proteins, Cancer, Cellular Markers, ER Markers, Hypoxia
Human,Mouse,Rat
Flow Cytometry,Western Blot
1 mg/mL
Western Blot 1:1000, Flow Cytometry 1:50-1:100
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
64714
Rabbit
IgG
Primary
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit