Learn More
PDI™ Sani-Cloth™ HB Germicidal Disposable Wipes
Alcohol free, quaternary formula
Manufacturer:  Nice Pak ProductsSupplier Diversity Partner Q85484
 View more versions of this product
Catalog No. 23-763-403
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EPA, CDC, OSHA
Canister
Large
Broad range of hard, non porous surfaces and equipment commonly found in healthcare environments, kills 99.999% of germs on many surfaces throughout fitness centers
5% Organic soil
Yes
10 min.
Safety and Handling
ShelfLife : 2 Year(s)