Learn More
PDI™ Sani-Cloth™ Plus Germicidal Disposable Cloths
Effective against 16 microorganisms in five minutes, including bloodborne pathogens, viruses and bacteria
Manufacturer:  Nice Pak ProductsSupplier Diversity Partner Q85084
 View more versions of this product
Catalog No. 06-669-55
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CDC, OSHA
Canister
65 wipes
19 × 38.1cm
3 min.
14.85% Quaternary/low-alcohol
Yes
7.5 × 15 in.
XL
Broad range of hard, non porous surfaces and equipment commonly found in healthcare environments
Safety and Handling
ShelfLife : 2 Year(s)