PDI™ Sani-Hands™ Instant Hand Sanitizing Wipes

Clean and sanitize hands when soap and water are not available

Manufacturer: Nice Pak ProductsSupplier Diversity Partner D43600

 View more versions of this product

Catalog No. 23-100-101

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Packaging Packets
    Quantity 100 Pack
    Certifications/Compliance FDA
    Composition Alcohol 65.9%
    For Use With (Application) Kills 99.9% of germs
    Format Wipe
    Product Type Hand Sanitizing Wipe

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit