Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical PDI™ Super Sani-Cloth™ Disposable Wipes

Kills a broad range of viruses and bacteria

Specifications

Color Clear
Product Type Super Sani-Cloth™ Germicidal Wipe
Format Pre-moistened wipe
For Use With (Application) Kills a broad range of viruses and bacteria TB, RSV, HBV, HIV, MRSA, VRE, Candida Albicans and Rhinovirus
View More Specs

Products 1
Catalog Number Mfr. No. Size (Metric) Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Size (English) Price Quantity & Availability  
19-898-564
View Documents
Moore Medical
76874
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Effective against 27 microorganisms including the following MDROs, bloodborne pathogens and viruses:

  • Acinetobacter baumannii
  • Klebsiella pneumoniae
  • Candida albicans
  • ESBL producing E. coli
  • Influenza A
  • MRSA
  • VRE
  • HIV
  • HBV
  • HCV

  • Compatible with a broad range of surfaces and equipment in healthcare
  • Bactericidal, Tuberculocidal, Virucidal

Compliance

Meets CDC, OSHA and CMS Tag F441 guidelines

Specifications

Specifications

Clear
Super Sani-Cloth™ Germicidal Wipe
Pre-moistened wipe
Kills a broad range of viruses and bacteria TB, RSV, HBV, HIV, MRSA, VRE, Candida Albicans and Rhinovirus
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit