Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pdi1p Mouse anti-Yeast, Clone: 38H8, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Pdi1p |
---|---|
Clone | 38H8 |
Concentration | 6.6mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Pdi1p Monoclonal specifically detects Pdi1p in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Pdi1p | |
6.6mg/mL | |
Unconjugated | |
Mouse | |
Yeast | |
Yeast nuclear preparations | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
38H8 | |
Monoclonal | |
Purified | |
RUO | |
MFP 1, MFP1, PDI, PDI 1, PDI1, Protein disulfide isomerase, Thioredoxin related glycoprotein 1, TRG 1, TRG1 | |
IgG1 | |
Tissue culture supernatant | |
58.2 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title