Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDIA4 Mouse anti-Human, Clone: OTI2B11, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503885
Description
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.Specifications
PDIA4 | |
Monoclonal | |
0.45 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P13667 | |
ERP70, ERP72, ERp-72 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
9601 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2B11 | |
Unconjugated | |
PDIA4 | |
Liquid | |
PDIA4 | |
Full length human recombit protein of human PDIA4 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title