Learn More
Aviva Systems Biology Corp PDIA6 Antibody - N-terminal region (ARP52102_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52102_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDIA6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PDIA6. Alternate names: ERP5, P5, TXNDC7ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60681