Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDILT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559782
Description
Immunogen sequence: VSSIHITKPV HILEERSLLV LTPAGLTQML NQTRFLMVLF HNPSSKQSRN LAEELGKAVE IMGKGKNGIG FGKVDITIEK ELQQEFGI Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 72%.
Probable redox-inactive chaperone involved in spermatogenesis.
Specifications
PDILT | |
Polyclonal | |
Unconjugated | |
PDILT | |
PDIA7; PDILT; protein disulfide isomerase family A, member 7; protein disulfide isomerase like, testis expressed; protein disulfide isomerase-like protein of the testis; protein disulfide isomerase-like, testis expressed; protein disulfide-isomerase-like protein of the testis | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
204474 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N807 | |
PDILT | |
Recombinant protein corresponding to Human PDILT. Recombinant protein control fragment (Product #RP-98764). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction