Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDK2 Mouse anti-Human, Mouse, Clone: 180CT10.2.3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524716

Catalog No. PIMA524716

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
Specifications

Specifications

PDK2
Monoclonal
0.38 mg/mL
PBS with 0.09% sodium azide
Q15119, Q9JK42
[Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial, Pyruvate dehydrogenase kinase isoform 2, PDH kinase 2, PDKII, PDK2, PDHK2
Mouse
IgG1, kappa
400 μL
-20°C
Primary
18604, 5164
Immunohistochemistry (Paraffin), Western Blot
180CT10.2.3
Unconjugated
PDK2
Liquid
PDK2
human PDK2 recombit protein
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit