Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDK3 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548677

Catalog No. PA548677

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
Specifications

Specifications

Pdk3
Polyclonal
PBS with 0.09% sodium azide
Q922H2
[Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial, Pyruvate dehydrogenase kinase isoform 3, Pdk3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
236900
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Pdk3
Liquid
PDK3
KLH conjugated synthetic peptide between 387-414 amino acids from the C-terminal region of mouse Pdk3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit