Learn More
Neuvitro PDL PRE COATED COVERSLIPS 22MM
Manufacturer: Neuvitro H22PDL
Pre-coated CE certified German glass coverslips with Poly-D-Lysine for cell culture and confocal / IHC imaging, 22mm diameter round, #1 thickness, 45 pieces, sterile, ready to use.