Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDLIM1 Mouse anti-Human, Clone: CPTC-PDLIM1-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30751810UG

 View more versions of this product

Catalog No. NB122509


Add to cart

Description

Description

PDLIM1 Monoclonal specifically detects PDLIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

PDLIM1
CPTC-PDLIM1-1
Unconjugated
10 mM PBS with 0.05% BSA
CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1
Recombinant human full-length PDLIM1 protein
Protein A or G purified
RUO
Primary
9124
Western Blot, Immunohistochemistry (Paraffin)
0.2 mg/ml
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit