Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDLIM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531151
Description
Recommended positive controls: A549, HeLa, HepG2. Predicted reactivity: Mouse (85%), Rat (83%), Pig (84%), Rabbit (87%), Rhesus Monkey (97%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein which may be involved in bone development. Mutations in this gene are associated with susceptibility to osteoporosis.
Specifications
PDLIM4 | |
Polyclonal | |
Unconjugated | |
PDLIM4 | |
enigma homolog; H-REV18; LIM domain protein; LIM protein RIL; Pdlim4; PDZ and LIM domain 4; PDZ and LIM domain protein 4; reversion induced LIM; reversion-induced LIM protein; RIL; RIT-18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P50479 | |
PDLIM4 | |
Recombinant protein fragment corresponding to a region within amino acids 2 and 228 of Human PDLIM4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction