Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDLIM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598042
Description
PDLIM4 Polyclonal Antibody for Western Blot, IP, ELISA
This gene encodes a protein which may be involved in bone development. Mutations in this gene are associated with susceptibility to osteoporosis.
Specifications
PDLIM4 | |
Polyclonal | |
Unconjugated | |
PDLIM4 | |
enigma homolog; H-REV18; LIM domain protein; LIM protein RIL; Pdlim4; PDZ and LIM domain 4; PDZ and LIM domain protein 4; reversion induced LIM; reversion-induced LIM protein; RIL; RIT-18 | |
Rabbit | |
Protein G | |
RUO | |
8572 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
3.05 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P50479 | |
PDLIM4 | |
Recombinant Human PDZ and LIM domain protein 4 protein (81-243AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction