Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDLIM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529394
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (88%), Rat (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.
Specifications
| PDLIM5 | |
| Polyclonal | |
| Unconjugated | |
| PDLIM5 | |
| 1110001A05Rik; AI987914; C87059; ENH; Enh1; Enh2; Enh3; Enigma homolog; enigma-like LIM domain protein; enigma-like PDZ and LIM domains protein; L9; LIM; Pdlim5; PDZ and LIM domain 5; PDZ and LIM domain protein 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96HC4 | |
| PDLIM5 | |
| Recombinant protein encompassing a sequence within the center region of human PDLIM5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction