Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | PDLIM5 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB125537
|
Novus Biologicals
NBP310318100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
PDLIM5 Polyclonal specifically detects PDLIM5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDLIM5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5 | |
The immunogen is a synthetic peptide directed towards the middle region of human PDLIM5 (NP_001011516). Peptide sequence VKIPPKRPPRKHIVERYTEFYHVPTHSDASKKRLIEDTEDWRPRTGTTQS | |
Affinity purified |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
10611 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title