Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549694

Catalog No. PA549694


Add to cart

Description

Description

The antibody detects endogenous level of total PDP1 protein.

Catalyzes the dephosphorylation and concomitant reactivation of the alpha subunit of the E1 component of the pyruvate dehydrogenase complex (Hu R.-M., Han Z.-G., Song H.-D., Peng Y.-D., Huang Q.-H., Ren S.-X., Gu Y.-J., Huang C.-H., Li Y.-B., Jiang C.-L., Fu G., Zhang Q.-H., Gu B.-W., Dai M., Mao Y.-F., Gao G.-F., Rong R., Ye M. Chen J.-L. Proc. Natl. Acad. Sci. U.S.A. 97:9543-9548(2000)).
Specifications

Specifications

PDP1
Polyclonal
Unconjugated
PDP1
Liquid
PDP1
Peptide sequence around aa.379∽383 (N-E-Y-T-K) derived from Human PDP1 .
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9P0J1, Q3UV70, O88483
PDP, PPM2C
Rabbit
IgG
100 μL
-20°C
Primary
381511, 54704, 54705
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit