Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDS5B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103601
Description
Antibody detects endogenous levels of total PDS5B.
This gene encodes a protein that interacts with the conserved protein complex termed cohesin. The cohesin complex holds together sister chromatids and facilitates accurate chromosome segregation during mitosis and meiosis. This protein is also a negative regulator of cell proliferation and may be a tumor-suppressor gene.
Specifications
PDS5B | |
Polyclonal | |
Unconjugated | |
PDS5B | |
AI646570; androgen induced inhibitor of proliferation; androgen-induced proliferation inhibitor; androgen-induced prostate proliferative shutoff associated protein; androgen-induced prostate proliferative shutoff associated protein AS3; androgen-induced prostate proliferative shutoff-associated protein AS3; androgen-induced shutoff 3; Aprin; As3; AW212954; CG008; Kiaa0979; mKIAA0979; PDS5 cohesin associated factor B; PDS5, regulator of cohesion maintenance, homolog B; PDS5, regulator of cohesion maintenance, homolog B (S. cerevisiae); Pds5b; Sister chromatid cohesion protein PDS5 homolog B | |
Rabbit | |
Affinity chromatography | |
RUO | |
100710, 23047, 304218 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q4VA53, Q6TRW4, Q9NTI5 | |
PDS5B | |
A synthesized peptide derived from human PDS5B(Accession Q9NTI5), corresponding to amino acid residues A1356-S1406. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction