Learn More
Selleck Chemical LLCSupplier Diversity Partner Pyrrolidinedithiocarbamate ammonium DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S363325MG
Ammonium pyrrolidine dithiocarbamate (PDTC) is a potent nuclear factor-κB (NF-κB) inhibitor that inhibits IκB phosphorylation, blocks NF-κB translocation to the nucleus and reduces the expression of downstream cytokines.
Catalog No. 50-194-8022