Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDX-1/IPF1 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: OTI2A12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273330V
Description
PDX-1/IPF1 Monoclonal specifically detects PDX-1/IPF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PDX-1/IPF1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PDX1 | |
Full length recombinant protein of human PDX-1/IPF1 (NP_000200) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2A12 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism, Stem Cell Markers | |
3651.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title