Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDX1 Mouse anti-Human, Clone:2G12, Abnova™
Mouse monoclonal antibody raised against partial recombinant human PDX1.
Supplier: Abnova Corporation MAB17873
Description
The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4). [provided by RefSeq]Specifications
PDX1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
PDX1 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
2G12 | |
pancreatic and duodenal homeobox 1 | |
In PBS (0.05% sodium azide) | |
GSF, IDX-1, IPF1, IUF1, MODY4, PDX-1, STF-1 | |
Recombinant protein corresponding to amino acids 39-283 of human PDX1. | |
3651 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction