Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDXP Polyclonal Antibody
PDXP Polyclonal Antibody
Supplier: Thermo Scientific PA544765
Description
PDXP Polyclonal Antibody for Western Blot

Specifications
PDXP | |
Polyclonal | |
Unconjugated | |
PDXP | |
1600027H05Rik; AB041662; chronophilin; chronophin; CIN; dJ37E16.5; FLJ32703; PDXP; Plp; PLP phosphatase; PLPP; PLP-phosophatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; pyridoxal phosphatase; pyridoxal phosphate phosphatase; Rbp1; reg I binding protein I; reg I-binding protein 1; testicular secretory protein Li 36 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57026 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96GD0 | |
PDXP | |
Synthetic peptide directed towards the middle region of human PDXP. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction