Learn More
Aat BioquestSupplier Diversity Partner PE [R-Phycoerythrin] *CAS 11016-17-4*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2556
R-Phycoerythrin (PE) is isolated from red algae. Its primary absorption peak is at 565 nm with secondary peaks at 496 and 545 nm. The relative prominence of the secondary peaks varies significantly among R-PEs from different species.
Catalog No. 50-195-3185