Learn More
BiolegendSupplier Diversity Partner PE anti-human CD229 (Ly-9) DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 326108
PE anti-human CD229 (Ly-9) [HLy-9.1.25]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-314