Learn More
BiolegendSupplier Diversity Partner PE anti-human MSC (W7C6) DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 327606
PE anti-human MSC (W7C6) [W7C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-167-381