Learn More
BiolegendSupplier Diversity Partner PE anti-human SSEA-4 DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 330405
PE anti-human SSEA-4 [MC-813-70]; Isotype: Mouse IgG3, κ; Reactivity: Human, Baboon, Cynomolgus, Mouse, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-568