Learn More
BiolegendSupplier Diversity Partner PE anti-mouse CD104  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 123610
PE anti-mouse CD104 [346-11A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-711-863