Learn More
BiolegendSupplier Diversity Partner PE/Cyanine7 anti-human CD303 (BDCA-2)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 354214
PE/Cyanine7 anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-143