Learn More
BiolegendSupplier Diversity Partner PE/Dazzle™ 594 anti-mouse/human CD11b  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 101256
PE/Dazzle™ 594 anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit (Lapine); Apps: FC; Size: 100 ug
Catalog No. 50-711-814