Learn More
Aat BioquestSupplier Diversity Partner ReadiUse™ Preactivated PE-iFluor™ 750 Tandem  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2578
R-Phycoerythrin (PE) is isolated from red algae. Its primary absorption peak is at 565 nm with secondary peaks at 496 and 545 nm.
Catalog No. 50-195-3193