Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peanut Oil, NF, Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle

PE090, 8002-03-7

Spectrum™ Peanut Oil, NF is used as a vehicle for pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Water 0.001
Quantity 500 mL
CAS 8002-03-7
CAS Max % 1
Grade NF