Learn More
R Biopharm Inc Lateral Flow Peanut Assay 25 determinations
Manufacturer:  R Biopharm Inc BL60625
Encompass
The Lateral Flow Peanut assay is used for the qualitative detection of peanut residues in environmental / surface samples (e.g. in food production lines). The assay is also applicable for food samples. The assay is applied for the detection of small amounts of peanut contaminations.
Catalog No. NC1857023