Learn More
Zyagen LabsSupplier Diversity Partner Pear total RNA is a high pure intact total RNA isolated from freshly harvested PEAR leaves, treated with DNase and dissolved in water. RNA is ready for immediate use in any downstream application. DFS Item
Manufacturer:  Zyagen LabsSupplier Diversity Partner PLR1033
Zyagen Total RNA is routinely extracted from freshly harvested plant leaves using classical guanidine isothiocyanate–phenol:chloroform extraction method which allows the rapid isolation of total RNA including microRNAs. RNA is treated with RNase-free DNase to remove residual DNA, precisely quantified, and stored at -80oC. The integrity of each RNA sample as indicated by intact ribosomal RNA is verified by denatured agarose gel electrophoresis. The purity of RNA is assessed by NanoDrop (A260/A280: 1.9-2.1). Residual DNA contamination is tested by PCR. RNA is ideal for Northern blotting, ribonuclease protection assay, SI nuclease assay, RT-PCR/Q-PCR/RACE analysis, cDNA synthesis, RNA differential display, microRNA studies, and purification of mRNA for library construction. Total RNA sample is provided in RNase-free water at a concentration of 1 mg/ml and shipped on dry ice.
Catalog No. 50-171-8448