Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEAR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112645
Description
PEAR1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Platelet endothelial aggregation receptor 1 (PEAR1) is a platelet receptor that signals upon the formation of platelet-platelet contacts independent of platelet activation and secondary to platelet aggregation. Upon platelet aggregation stimulated by physiological agonists, PEAR1 becomes tyrosine- and serine-phosphorylated; the tyrosine phosphorylation can be inhibited by eptifibatide, an aIIbbeta3 antagonist that also inhibits platelet aggregation. Recent studies have indicated that genetic variations in PEAR1, may be associated with enhanced agonist-induced platelet aggregation.
Specifications
PEAR1 | |
Polyclonal | |
Unconjugated | |
PEAR1 | |
3110045G13Rik; hPEAR1; Jagged and Delta protein; Jedi; Jedi-1; MEGF12; mPEAR1; Multiple EGF-like domains protein 12; multiple EGF-like-domains 12; multiple epidermal growth factor-like domains protein 12; Pear1; Platelet endothelial aggregation receptor 1; protein Jedi; RGD1305653; RP11-107D16.3 | |
Rabbit | |
Protein G | |
RUO | |
295293, 375033, 73182 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q5VY43, Q8VIK5 | |
PEAR1 | |
Recombinant Human Platelet endothelial aggregation receptor 1 protein (932-1037AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction