Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1 Mouse anti-Human, Clone: C31.10, Abnova™

Mouse monoclonal antibody raised against full length recombinant human PECAM1.

Manufacturer:  Abnova Corporation MAB13395

Catalog No. 89-934-753


Add to cart

Description

Description

Specifications

Specifications

PECAM1
C31.10
Mouse monoclonal antibody raised against full length recombinant human PECAM1.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
CD31/PECAM-1
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
PECAM1
Recombinant protein corresponding to full length human PECAM1.
Protein A/G purification
RUO
Primary
5175
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit