Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PECAM1 Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA505003
Description
CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. The N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. Further, CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. CD31 is one of key regulatory molecules in the vascular system. CD31 induces susceptibility to atherosclerosis and this cell adhesion molecule is required for leukocyte migration under most inflammatory conditions. Antibodies to CD31 is of value in the study of benign and maligt vascular tumors.Specifications
PECAM1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P16284 | |
PECAM1 | |
Full length human recombit protein of human PECAM1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2C3 | |
Unconjugated | |
PECAM1 | |
CD31, CD31/EndoCAM, GPIIA′, PECA1, PECAM-1, endoCAM | |
Mouse | |
Affinity Chromatography | |
RUO | |
5175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction