Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1 Mouse anti-Human, Clone: TP1/15 (OC-515), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13874

Catalog No. 89-033-939

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mouse monoclonal antibody raised against native human PECAM1.

  • High specificity
  • High reactivity
Specifications

Specifications

PECAM1
TP1/15
Mouse monoclonal antibody raised against native human PECAM1.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
PECAM1
Native purified PECAM1 from human myeloid leukemia RC-2A cell line.
Protein A/G purification
RUO
Primary
5175
Flow Cytometry
Unconjugated
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
P16284
CD31|PECAM-1
Mouse
IgG2a
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)