Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PECAM1 Mouse anti-Human, PE, Clone: SPM122, Abnova™
Mouse monoclonal antibody raised against native human PECAM1.
Supplier: Abnova Corporation MAB13016
Specifications
PECAM1 | |
Monoclonal | |
PE | |
Flow Cytometry (5 uL per test per 1 million cells in 0.1 mL) Immunofluorescence (1:50-1:100) The optimal working dilution should be determined by the end user. | |
CD31/PECAM-1 | |
Mouse | |
Protein A/G purification | |
RUO | |
5175 | |
Store at 4°C. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
SPM122 | |
Mouse monoclonal antibody raised against native human PECAM1. | |
In PBS (0.05% BSA, 0.05% sodium azide). | |
PECAM1 | |
Membrane preparation of a spleen from a patient with hairy cell leukemia. | |
500 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction