Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PECI Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532008
Description
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PECI is an auxiliary enzyme that catalyzes an isomerization step required for the beta-oxidation of unsaturated fatty acids.
Specifications
PECI | |
Polyclonal | |
Unconjugated | |
ECI2 | |
ACBD2; acyl-Coenzyme A binding domain containing 2; D3,D2-enoyl-CoA isomerase; DBI-related protein 1; delta(3),delta(2)-enoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; dJ1013A10.3; Dodecenoyl-CoA isomerase; Drs1; DRS-1; Eci2; enoyl-CoA delta isomerase 2; enoyl-CoA delta isomerase 2, mitochondrial; enoyl-Coenzyme A delta isomerase 2; Hca88; Hepatocellular carcinoma-associated antigen 88; PECI; Peroxisomal 3,2-trans-enoyl-CoA isomerase; peroxisomal 3,2-trans-enoyl-CoA isomerase; enoyl-CoA delta isomerase 2, mitochondrial; peroxisomal D3,D2-enoyl-CoA isomerase; peroxisomal delta3, delta2-enoyl-Coenzyme A isomerase; renal carcinoma antigen NY-REN-1; testicular secretory protein Li 33 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10455 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.93 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75521 | |
ECI2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 307 of Human PECI. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction