Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PECR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112329
Description
PECR Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.
Specifications
PECR | |
Polyclonal | |
Unconjugated | |
Pecr | |
2,4-dienoyl-CoA reductase-related protein; 2400003B18Rik; DCRRP; DCR-RP; HPDHase; HSA250303; PECR; perosisomal 2-enoyl-CoA reductase; peroxisomal 2,4-dienoyl-CoA reductase; Peroxisomal trans-2-enoyl-CoA reductase; PRO1004; putative short chain alcohol dehydrogenase; PVIARL; pVI-ARL; PX-2,4-DCR1; RLF98; SDR29C1; Short chain dehydrogenase/reductase family 29C member 1; short chain dehydrogenase/reductase family 29C, member 1; TERP | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
55825 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9BY49 | |
Pecr | |
E. coli-derived Human PECR fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction