Learn More
Thermo Scientific Chemicals Pectinase, Aspergillus niger
Used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.
$158.00 - $489.65
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ63408MD
|
Thermo Scientific Chemicals
J63408MD |
250 mg |
Each for $158.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ6340803
|
Thermo Scientific Chemicals
J6340803 |
1 g |
Each
for $489.65
Save 5% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Powder |
Safety and Handling
EINECSNumber : 232-885-6
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.