Learn More
Sklar InstrumentsSupplier Diversity Partner Pederson Vaginal Speculum, Sklar, 1/2 in. x 3 in., Straight, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 903730
Pederson Vaginal Speculum, Sklar, 1/2 in. x 3 in., Straight, Stainless Steel. The Sklar Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient. This product is a small sized speculum with 3 in. X 1/2 in. blades. It features gold external screws.
Catalog No. 50-143-6665