Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEDF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA586509
Description
The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.Specifications
PEDF | |
Polyclonal | |
Unconjugated | |
SERPINF1 | |
Cell proliferation-inducing gene 35 protein; EPC 1; EPC-1; EPC1; PEDF; PEDF; PIG 35; PIG35; Pigment epithelium derived factor; Pigment epithelium-derived factor; Proliferation inducing protein 35; Serpin F1; Serpin peptidase inhibitor clade F member 1; SERPINF 1; Serpinf1; | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20317, 287526, 5176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P36955, P97298 | |
Serpinf1 | |
Synthetic peptide, corresponding to amino acids 280-320 of Human PEDF. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title