Learn More
Thermo Scientific™ PEEK Capillary Tubing Available on GSA/VA Contract for Federal Government customers only.
Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors. 
Manufacturer:  Thermo Scientific™ 3700520
 View more versions of this product
Catalog No. 03-050-107
  Check Availability
Add to cart
Description
Description
Use Thermo Scientific™ Rheodyne™ Sample Loops  for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.
  • Broad chemical compatibility
  • Biocompatible
  • Easily cut to desired length
  • Appropriate for many HPLC applications
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.005 in. ID; 20cm length; Red
Rheodyne™ HPLC sample injectors
Capillary Tubing
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.